Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MHC I (HLA-A, B, C) Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in flow cytometry experiments with the Thermo Scientific™ MHC I (HLA-A, B, C) Ab-1, Mouse Monoclonal Antibody.

$238.04 - $819.63

Specifications

Antigen MHC I (HLA-A, B, C) Ab-1
Clone 246-B8.E7
Applications Flow Cytometry
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS557P

 
thermo scientific™ lab vision™
MS557P
200μg/mL 1mL Each for $724.35

MS557P0

 
thermo scientific™ lab vision™
MS557P0
200μg/mL 100μL Each for $238.04

MS557P1

 
thermo scientific™ lab vision™
MS557P1
200μg/mL 500μL Each for $574.33

MS557P1ABX

 
thermo scientific™ lab vision™
MS557P1ABX
1mg/mL 100μL Each for $649.14

MS557PABX

 
thermo scientific™ lab vision™
MS557PABX
1mg/mL 200μL Each for $819.63
Description & Specifications

Specifications

Antigen MHC I (HLA-A, B, C) Ab-1
Clone 246-B8.E7
Applications Flow Cytometry
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Non-Human Primate
Cross Reactivity Human
Cross Reactivity Non-Human Primate
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Hematopoietic
Type Primary
Immunogen Human peripheral blood leukocytes
Isotype IgG2a kappa

Ensure accurate, reproducible results in flow cytometry experiments with the Thermo Scientific™ MHC I (HLA-A, B, C) Ab-1, Mouse Monoclonal Antibody.

Human MHC class I antigens (HLA-A, B and C) are expressed constitutively on all nucleated cells lymphocytes such as lymphocytes, thymocytes, granulocytes, and bone marrow cells and are absent on erythrocytes. MHC class I antigens play a role in class I MHC- associated antigen presentation, inhibition of NK cell cytotoxicity, tumor surveillance, and tissue allotransplantation.

Host Species: Mouse

Clone: 246-B8.E7

Isotype: IgG2a/κ

Species Reactivity: Human and Primates. Others not known.

Epitope: Not determined

Immunogen: Human peripheral blood leukocytes

Positive Control: Human lymphocytes, Tonsil and lymph node

Cellular Localization: Membrane

Recommended for:

  • Flow cytometry

Note:

USA: RUO; Int'l: RUO