Please login to your online account to display your discounted pricing

MHC Class II Rat anti-Canine, FITC, Clone: YKIX334.2, eBioscience™

Rat Monoclonal Antibody

$143.55 - $368.30

Specifications

Antigen MHC Class II
Clone YKIX334.2
Host Species Rat
Species Reactivity Canine
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122149

 
life technologies
LS11590942
100 Tests Each for $368.30

501122148

 
life technologies
LS11590941
25 Tests Each for $143.55
Description & Specifications

Specifications

Antigen MHC Class II
Clone YKIX334.2
Host Species Rat
Species Reactivity Canine
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

This YKIX334.2 monoclonal antibody reacts with canine major histocompatibility complex (MHC) Class II. The MHC plays a significant role in immunity by presenting self and non-self antigens to T cells. In dogs, MHC Class II is expressed on all T and B cells, macrophages, fibroblasts, and kidney interstitial cells.

This monoclonal antibody has been reported to inhibit mixed lymphocyte reactions and T cell activation induced by MHC Class II antigen presentation in vitro. Also, YKIX334.2 recognition of MHC Class II is not allele-specific.