missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MHC class II (I-Ek) Mouse anti-Mouse, Rat, PE, Clone: 14-4-4S, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $230.55

Specifications

Antigen MHC class II (I-Ek)
Clone 14-4-4S
Host Species Murine
Gene Alias MHC II, IE-k, IEk
Species Reactivity Mouse, Rat
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011045 Life Technologies
LS12598081
50μg Each for $143.55
5011046 Life Technologies
LS12598082
100μg Each for $230.55
Specifications

Description

The 14-4-4S monoclonal antibody reacts with the mouse and rat major histocompatibility complex class II, I-E subregion-encoded glycoprotein in mice strains of H-2k, H-2d, H-2p and H-2r. H-2b, H-2f and H-2s haplotypes do not express the I-E antigen. MHC class II is present on B cells, monocytes, macrophages, and dendritic cells. The 14-4-4S mAb has been reported to block antigen presentation in in vitro assays. This antibody cross-reacts with rat.
Description & Specifications

Specifications

MHC class II (I-Ek)
Murine
Mouse, Rat
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
14-4-4S
MHC II, IE-k, IEk
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL