missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MHC Class II (I-A/I-E), PE-eFluor 610, clone: M5/114.15.2, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS61532180

 View more versions of this product

Catalog No. 501124831



Description

Description

MHC (major histocompatibility complex) class II molecules are transmembrane glycoproteins expressed on the surface of professional antigen-presenting cells, such as macrophages, dendritic cells and B cells. Before their exposition on the cell surface, the MHC class II molecules react with endocytosed exogenous antigens, which are then presented to the T cells. The antigen-binding grove between MHC class II alpha and beta chain is open at both ends and is 15-24 amino acid residues long.

Please make sure that your instrument is capable of detecting this fluorochome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. MHC (major histocompatibility complex) class II molecules are transmembrane glycoproteins expressed on the surface of professional antigen-presenting cells, such as macrophages, dendritic cells and B cells. Before their exposition on the cell surface, the MHC class II molecules react with endocytosed exogenous antigens, which are then presented to the T cells. The antigen-binding grove between MHC class II alpha and beta chain is open at both ends and is 15-24 amino acid residues long.
Specifications

Specifications

MHC Class II (I-A/I-E)
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P04231, P06342
I-Ab, I-Ad, I-Aq, I-Ed, I-Ek, MHC class 2, MHC class II glygoproteins
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
M5/114.15.2
PE-eFluor® 610
H2-Ab1
Liquid
H2-Ab1, H2-Eb1
IgG2b, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
14961, 14969
Documents

For Research Use Only.