Please login to your online account to display your discounted pricing

MHC Class II (I-A/I-E) Rat anti-Mouse, eVolve 655, Clone: M5/114.15.2, eBioscience™

Rat Monoclonal Antibody

$172.55 - $397.30

Specifications

Antigen MHC Class II (I-A/I-E)
Clone M5/114.15.2
Host Species Rat
Gene Alias MHC II, IA, IE, I-A/E, IA/IE
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125052

 
life technologies
LS86532141
25 tests Each for $172.55

501125053

 
life technologies
LS86532142
100 tests Each for $397.30
Description & Specifications

Specifications

Antigen MHC Class II (I-A/I-E)
Clone M5/114.15.2
Host Species Rat
Gene Alias MHC II, IA, IE, I-A/E, IA/IE
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eVolve 655
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This product should be used within 6 months from the date of receipt.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL

The M5/114.15.2 monoclonal antibody reacts with the mouse major histocompatibility complex class II, both I-A and I-E subregion-encoded glycoproteins (I-A b, I-A d, I-A q, I-E d, I-E k, not I-A f, I-A k, or I-A s). It detects a polymorphic determinant present on B cells, monocytes, macrophages, dendritic cells, and activated T lymphocytes from mice carrying the H-2 b, H-2 d, H-2 q, H-2 p, H-2 r and H-2 u but not from mice carrying the H-2 s or H-2 f haplotypes. The M5/114 mAb is reported to inhibit I-A-restricted T cell responses of the H-2 b, H-2 d, H-2 q, H-2 u but not H-2 f, H-2 k, or H-2 s haplotypes.