missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MHC Class II (I-A/I-E) Rat anti-Mouse, Clone: M5/114.15.2, eBioscience™

Rat Monoclonal Antibody

$91.08 - $315.48

Specifications

Antigen MHC Class II (I-A/I-E)
Clone M5/114.15.2
Host Species Rat
Gene Alias MHC II, IA, IE, I-A/E, IA/IE
Applications Flow Cytometry,Functional Assay,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013090 Life Technologies
LS14532181
50μg Each for $91.08
501129377 Life Technologies
LS14532182
100μg Each for $143.88
501129455 Life Technologies
LS14532185
500μg Each for $315.48
Specifications

Description

The M5/114.15.2 monoclonal antibody reacts with the mouse major histocompatibility complex class II, both I-A and I-E subregion-encoded glycoproteins (I-A b, I-A d, I-A q, I-E d, I-E k, not I-A f, I-A k, or I-A s). It detects a polymorphic determinant present on B cells, monocytes, macrophages, dendritic cells, and activated T lymphocytes from mice carrying the H-2 b, H-2 d, H-2 q, H-2 p, H-2 r and H-2 u but not from mice carrying the H-2 s or H-2 f haplotypes. The M5/114 mAb is reported to inhibit I-A-restricted T cell responses of the H-2 b, H-2 d, H-2 q, H-2 u but not H-2 f, H-2 k, or H-2 s haplotypes.
Description & Specifications

Specifications

MHC Class II (I-A/I-E)
Rat
Flow Cytometry,Functional Assay,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
M5/114.15.2
MHC II, IA, IE, I-A/E, IA/IE
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer