missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MHC Class II (I-A/I-E) Rat anti-Mouse, APC, Clone: M5/114.15.2, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS17532181

 View more versions of this product

Catalog No. 501129473



Description

Description

Description: The M5/114.15.2 monoclonal antibody reacts with the mouse major histocompatibility complex class II, both I-A and I-E subregion-encoded glycoproteins (I-A b, I-A d, I-A q, I-E d, I-E k, not I-A f, I-A k, or I-A s). It detects a polymorphic determit present on B cells, monocytes, macrophages, dendritic cells, and activated T lymphocytes from mice carrying the H-2 b, H-2 d, H-2 q, H-2 p, H-2 r and H-2 u but not from mice carrying the H-2 s or H-2 f haplotypes. The M5/114 mAb is reported to inhibit I-A-restricted T cell responses of the H-2 b, H-2 d, H-2 q, H-2 u but not H-2 f, H-2 k, or H-2 s haplotypes. Applications Reported: M5/114.15.2 has been reported for use in flow cytometric analysis. Applications Tested: The M5/114.15.2 antibody has been tested by flow cytometric analysis of mouse splenocytes and can be used at less than or equal to 0.03 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

MHC (major histocompatibility complex) class II molecules are transmembrane glycoproteins expressed on the surface of professional antigen-presenting cells, such as macrophages, dendritic cells and B cells. Before their exposition on the cell surface, the MHC class II molecules react with endocytosed exogenous antigens, which are then presented to the T cells. The antigen-binding grove between MHC class II alpha and beta chain is open at both ends and is 15-24 amino acid residues long.
Specifications

Specifications

MHC Class II (I-A/I-E)
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P04231, P06342
I-Ab, I-Ad, I-Aq, I-Ed, I-Ek, MHC class 2, MHC class II glygoproteins
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
M5/114.15.2
APC
H2-Ab1
Liquid
H2-Ab1, H2-Eb1
IgG2b, kappa
50 μg
4° C, store in dark, DO NOT FREEZE!
Primary
14961, 14969
Documents

For Research Use Only.