Please login to your online account to display your discounted pricing

MHC Class I (RT1-A) Mouse anti-Rat, FITC, Clone: OX18, eBioscience™

Mouse Monoclonal Antibody

$274.05 - $274.05

Specifications

Antigen MHC Class I (RT1-A)
Clone OX18
Host Species Murine
Species Reactivity Rat
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122082

 
life technologies
LS11092182
100μg Each for $274.05
Description & Specifications

Specifications

Antigen MHC Class I (RT1-A)
Clone OX18
Host Species Murine
Species Reactivity Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

This OX18 monoclonal antibody recognizes a non-polymorphic determinant of rat MHC Class I, which is also known as RT1-A. Rat MHC Class I is expressed on all nucleated cells and erythrocytes.

The OX18 antibody has been reported to have functional activity.