Please login to your online account to display your discounted pricing

MHC Class I (H-2Kk) Mouse anti-Mouse, eFluor 450, Clone: AF3-12.1.3, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $288.55

Specifications

Antigen MHC Class I (H-2Kk)
Clone AF3-12.1.3
Host Species Mouse
Gene Alias MHC I, H2Kk, H2-Kk, H2K-k
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016385

 
affymetrix
48594080
25μg Each for $129.05

5016386

 
affymetrix
48594082
100μg Each for $288.55
Description & Specifications

Specifications

Antigen MHC Class I (H-2Kk)
Clone AF3-12.1.3
Host Species Mouse
Gene Alias MHC I, H2Kk, H2-Kk, H2K-k
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This AF3-12.1.3 monoclonal antibody reacts with the H-2Kk MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8. Reactivity to other haplotypes (e.g. b, f, j, k, p, q, r, s, u, and v) has not been observed.