Please login to your online account to display your discounted pricing

MHC Class I (H-2Kd) Mouse anti-Mouse, eFluor 450, Clone: SF1-1.1.1, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $361.05

Specifications

Antigen MHC Class I (H-2Kd)
Clone SF1-1.1.1
Host Species Murine
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50246023

 
life technologies
LS48595780
25μg Each for $143.55

50246024

 
life technologies
LS48595782
100μg Each for $361.05
Description & Specifications

Specifications

Antigen MHC Class I (H-2Kd)
Clone SF1-1.1.1
Host Species Murine
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This SF1-1.1.1 monoclonal antibody reacts with the H-2Kd MHC class I alloantigen. H-2Kd is involved in antigen presentation to T cells expressing CD3/TCR and CD8. Reactivity to other haplotypes (e.g. b, j, k, p, q, s, and v) has not been observed.