Please login to your online account to display your discounted pricing

MHC Class I (H-2Kd) Mouse anti-Mouse, Biotin, Clone: SF1-1.1.1, eBioscience™

Mouse Monoclonal Antibody

$56.55 - $187.05

Specifications

Antigen MHC Class I (H-2Kd)
Clone SF1-1.1.1
Host Species Murine
Gene Alias MHCI
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012062

 
life technologies
LS13595780
25μg Each for $56.55

5012063

 
life technologies
LS13595782
100μg Each for $187.05
Description & Specifications

Specifications

Antigen MHC Class I (H-2Kd)
Clone SF1-1.1.1
Host Species Murine
Gene Alias MHCI
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

This SF1-1.1.1 monoclonal antibody reacts with the H-2Kd MHC class I alloantigen. H-2Kd is involved in antigen presentation to T cells expressing CD3/TCR and CD8. Reactivity to other haplotypes (e.g. b, j, k, p, q, s, and v) has not been observed.