Please login to your online account to display your discounted pricing

MHC Class I (H-2Kd/H-2Dd) Mouse anti-Mouse, PerCP-eFluor 710, Clone: 34-1-2S, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $332.05

Specifications

Antigen MHC Class I (H-2Kd/H-2Dd)
Clone 34-1-2S
Host Species Mouse
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123879

 
life technologies
LS46599880
25μg Each for $129.05

501123880

 
life technologies
LS46599882
100μg Each for $332.05
Description & Specifications

Specifications

Antigen MHC Class I (H-2Kd/H-2Dd)
Clone 34-1-2S
Host Species Mouse
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 34-1-2S monoclonal antibody reacts with the mouse MHC class I, H-2Kd and H-2Dd. This cytotoxic antibody also cross-reacts with Kb, s, r, q, p.