missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MHC Class I (H-2Kd/H-2Dd) Mouse anti-Mouse, FITC, Clone: 34-1-2S, eBioscience™

Mouse Monoclonal Antibody

Manufacturer:  Life Technologies LS11599881

 View more versions of this product

Catalog No. 501128882



Description

Description

Description: The 34-1-2S monoclonal antibody reacts with the mouse MHC class I, H-2Kd and H-2Dd. This cytotoxic antibody also cross-reacts with Kb, s, r, q, p. Applications Reported: 34-1-2S has been reported for use in flow cytometric analysis. Applications Tested: This 34-1-2S antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

This protein is an MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8.
Specifications

Specifications

MHC Class I (H-2Kd/H-2Dd)
Monoclonal
0.5 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P01902, P14426
H-2 class I histocompatibility antigen, K-D alpha chain, H2K1, H2KD, H2DD
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
34-1-2S
FITC
H2-D1
Liquid
H2-D1, H2-K1
IgG2a, kappa
50 μg
4° C, store in dark, DO NOT FREEZE!
Primary
14964, 14972
Documents

For Research Use Only.