missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MHC Class I (H-2Kd/H-2Dd) Mouse anti-Mouse, eFluor 450, Clone: 34-1-2S, eBioscience™

Mouse Monoclonal Antibody

$130.68 - $315.48

Specifications

Antigen MHC Class I (H-2Kd/H-2Dd)
Clone 34-1-2S
Host Species Murine
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124189 Life Technologies
LS48599880
25μg Each for $130.68
501124190 Life Technologies
LS48599882
100μg Each for $315.48
Specifications

Description

The 34-1-2S monoclonal antibody reacts with the mouse MHC class I, H-2Kd and H-2Dd. This cytotoxic antibody also cross-reacts with Kb, s, r, q, p.
Description & Specifications

Specifications

MHC Class I (H-2Kd/H-2Dd)
Murine
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
34-1-2S
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer