Please login to your online account to display your discounted pricing

MHC Class I free chain without B2 microglobulin Mouse anti-Human, APC, Clone: A4, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $332.05

Specifications

Antigen MHC Class I free chain without B2 microglobulin
Clone A4
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123243

 
life technologies
LS17995841
25 Tests Each for $129.05

501123244

 
life technologies
LS17995842
100 Tests Each for $332.05
Description & Specifications

Specifications

Antigen MHC Class I free chain without B2 microglobulin
Clone A4
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Allophycocyanin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The A4 monoclonal antibody reacts with the class I human leukocyte antigen (HLA) complex in the absence of β2 microglobulin. There are three class I α-chain genes in humans, called HLA-A, -B and -C. Normally, HLA class I proteins are associated non-covalently with β2 microglobulin (β2M). The function of the HLA class I complex is to present peptides derived from intracellular pathogens, including viral pathogens, on the cell surface to antigen specific CD8+ cytotoxic T cells. Class I HLA is expressed by the majority of nucleated cells.