Sartorius MGG Grade Glass Microfiber Filters w/o Binder
Binder free glass microfiber filters are suitable for analytical and gravimetric analyses. They are also used as prefilter.
Supplier:  Sartorius FT31106050
 View more versions of this product
Catalog No. 14-555-128
Description
Description
Ideal for critical analyses in which impurities can be washed out of the filter.
Specifications
Specifications
32mbar
Disc
1.5 μm
38 sec.
MGG
500°C
0.28 mm
5 cm
White
Borosilicate Glass Microfiber
Non-sterile
Ideal for critical analyses in which impurities can be washed out of the filter
932°F
0.01 in.
1.96 in.