MG 624, Tocris
α7 neuronal nicotinic receptor antagonist
Manufacturer:  R & D Systems RDS135650
 View more versions of this product
Catalog No. 13-565-0
Specifications Description & Specifications
Specifications
MG 624
77257-42-2
C22H30NOI
N, N, N-Triethyl-2-[4-(2-phenylethenyl)phenoxy]ethanaminium iodide
50mg
>98%
451.39
Chemical Identifiers
>98%
451.39
MG 624
C22H30NOI
N, N, N-Triethyl-2-[4-(2-phenylethenyl)phenoxy]ethanaminium iodide
Safety and Handling
Recommended Storage : Store at Room Temperature