Please login to your online account to display your discounted pricing

MG 624, Tocris

α7 neuronal nicotinic receptor antagonist

$177.37 - $755.54

Chemical Identifiers

Chemical Name or Material MG 624
Assay Percent Range >98%
Molecular Formula C22H30NOI
Formula Weight 451.39
Synonym N, N, N-Triethyl-2-[4-(2-phenylethenyl)phenoxy]ethanaminium iodide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

13-561-0

 
r & d systems
RDS135610
10mg Each for $177.37

13-565-0

 
r & d systems
RDS135650
50mg Each for $755.54
Description & Specifications

Specifications

Formula Weight 451.39
Quantity 10mg
Chemical Name or Material MG 624
Assay Percent Range >98%
Molecular Formula C22H30NOI
CAS 77257-42-2
Synonym N, N, N-Triethyl-2-[4-(2-phenylethenyl)phenoxy]ethanaminium iodide
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature