Please login to your online account to display your discounted pricing

MFZ 10-7, Tocris™

Negative allosteric modulator at mGlu5

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material MFZ 10-7
Assay Percent Range >99%
Molecular Formula C15H9FN2·HCl
Formula Weight 272.7
Synonym 3-Fluoro-5-[2-(6-methyl-2-pyridinyl)ethynyl]benzonitrile hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

443810

 
r & d systems
443810
10mg Each for $221.01

443850

 
r & d systems
443850
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 272.7
Quantity 10mg
CAS 1224431-15-5
Molecular Formula C15H9FN2·HCl
Assay Percent Range >99%
Synonym 3-Fluoro-5-[2-(6-methyl-2-pyridinyl)ethynyl]benzonitrile hydrochloride
Chemical Name or Material MFZ 10-7
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C