MFG Tray Chemical-Resistant Multipurpose Trays and Pans
Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
Manufacturer:  MFG Tray 6240085269
 View more versions of this product
Catalog No. 15-239-14
Description
Description
  • Reinforced white thermoset polyester composite construction Resistant to most detergents, mild acid and alkaline solutions, corrosive salts and many reagents
  • Withstand temperatures from -51° to +121°C (-60° to +250°F) continuous exposure, or 149°C (300°F) intermittent
  • High load-bearing capacity
  • Excellent dimensional stability Easily steam cleaned
Order Info
Available in 1 in. deep (2.5cm) trays, shallow pans from 1 1/4 to 2 3/4 in. deep (3 to 7cm), and ventilated stacking pans from 2 3/4 to 4 1/8 in. deep (7 to 10.5cm).
Specifications
Specifications
White
Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
-51° to 121°C
7cm
63.5cm
Multipurpose Tray and Shallow Pan
63.5cm
31 x 25 x 2.75 in. (63.5 x 63.5 x 7cm)
-60° to 250°F
2.75 in.
31 in.
Fiberglass
25 in.
Documents