MFG Tray Chemical-Resistant Multipurpose Trays and Pans
Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
Manufacturer:  MFG Tray 6100031537
 View more versions of this product
Catalog No. 15-239-12
Description
Description
  • Reinforced white thermoset polyester composite construction Resistant to most detergents, mild acid and alkaline solutions, corrosive salts and many reagents
  • Withstand temperatures from -51° to +121°C (-60° to +250°F) continuous exposure, or 149°C (300°F) intermittent
  • High load-bearing capacity
  • Excellent dimensional stability Easily steam cleaned
Order Info
Available in 1 in. deep (2.5cm) trays, shallow pans from 1 1/4 to 2 3/4 in. deep (3 to 7cm), and ventilated stacking pans from 2 3/4 to 4 1/8 in. deep (7 to 10.5cm).
Specifications
Specifications
White
Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
-51° to 121°C
4cm
93cm
Multipurpose Tray and Shallow Pan
44.5cm
36.5 x 17.5 x 1.6 in. (93 x 44.5 x 4cm)
-60° to 250°F
1.6 in.
36.5 in.
Fiberglass
17.5 in.
Documents