Please login to your online account to display your discounted pricing

Mevastatin, Tocris

HMG-CoA reductase inhibitor

$139.08 - $581.87

Chemical Identifiers

Chemical Name or Material Mevastatin
Assay Percent Range >98%
Molecular Formula C23H34O5
Formula Weight 390.52
Synonym (2S)-2-Methyl-(1S,7S,8S,8aR)-1,2,3,7,8,8a-hexahydro-7-methyl-8-[2-[(2R,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl butanoate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

152650

 
r & d systems
RDS152650
50mg Each for $581.87

152610

 
r & d systems
RDS152610
10mg Each for $139.08
Description & Specifications

Specifications

Formula Weight 390.52
Quantity 50mg
Chemical Name or Material Mevastatin
Synonym (2S)-2-Methyl-(1S,7S,8S,8aR)-1,2,3,7,8,8a-hexahydro-7-methyl-8-[2-[(2R,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl butanoate
Molecular Formula C23H34O5
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
CAS 73573-88-3
Safety and Handling

Store at Room Temperature