Please login to your online account to display your discounted pricing
Mettler Toledo™ Temperature Calibration Kits
For use with halogen moisture analyzers. Mettler Toledo Temperature Calibration Kits are used to adjust halogen drying units to ensure consistent results.
Manufacturer: mettler toledo™  ME63064204
 View more versions of this product
Catalog No. 01-909-385A
  • / Each
Description & Specifications
Specifications
Product Type Temperature Calibration Kit
For Use With (Equipment) Halogen Moisture Analyzer
Includes NIST certificate
Type Temperature calibration kit
  • Adjustment disc with precision thermometer
  • Sample pan handler