missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mettler Toledo™ SG8 SevenGo pro™ pH/Ion Meter
Built-in data memory and automatic functions for outstanding performance
Manufacturer: Mettler Toledo™ 51302581
Description
- Portable meter measures ion concentration, pH, mV and temperature
- Designed for easy, comfortable right- or left-handed operation
- Choice of three packaging configurations
- Waterproof and dustproof to IP-67 standard
- Large backlit display of measured value and settings
- Rotatable electrode clip that mounts left or right for single-handed operation
- IP-67 pH/Ion meter with expanded range for the highest demands
- Continuous indication of electrode and battery status
- Automatic endpoint and buffer recognition
- Automatic temperature compensation
- Three-point calibration with predefined or user-defined buffers
- Segmented or linear calibration as desired
- Five-point ion calibration with choice of 6 predefined and one user-defined buffer set
- Record date, time, time interval
- Intuitive menu operation using softkeys
- GLP memory for 200 datapoints
- Contact-free infrared communication with notebook, printer or PC
- ON/OFF backlit display
Order Info
Packaging Options
All packages include wrist strap, four AA batteries, operating instructions, test certificate, and declaration of conformity:
- Meter Only, without electrode
- Meter with InLab™ 413-SG electrode and 1.8m cable
Specifications
±0.01 | |
200 | |
Measures ion concentration, pH, mV, temp | |
21.9 cm | |
BNC (IP67), Cinch (IP67), NTC 30kilohm | |
4.5 cm | |
0 to 9999ppm | |
±0.2°C | |
0.71 lb. | |
3.5 in. | |
LCD | |
0 to 1999 mV | |
3/4 Digit |
5 | |
1.75 x 3.5 x 8.62 in. (4.5 x 8.89 x 21.9cm) | |
8.62 in. | |
pH/Ion Meter, four AA 1.5V batteries/1.3V NiMH accumulator | |
1.75 in. | |
pH/Ion Meter | |
0.1mV | |
0.1°C | |
325 g | |
8.89 cm | |
IR to printer or PC via RS232 or USB | |
-2 to 20 | |
0.1/0.01/0.001 |