Please login to your online account to display your discounted pricing

Mettler Toledo™ SG8 SevenGo pro™ pH/Ion Meter

Built-in data memory and automatic functions for outstanding performance

$1,599.30 - $2,113.19

Specifications

Accuracy (pH) ±0.01
Accuracy (Temperature) ±0.2°C
Applications Measures ion concentration, pH, mV, temp
Calibration Points (pH) 5
Data Points 200
View More Specs
Products
Catalog Number Mfr. No. Includes Product Type Price Quantity    

01-910-518

 
mettler toledo™
ME51302581
pH/Ion Meter, four AA 1.5V batteries/1.3V NiMH accumulator pH/Ion Meter Each for $1,599.30

01-910-519

 
mettler toledo™
ME51302582
pH/Ion Meter, InLab 413 SG electrode (1.8m cable), four AA 1.5V batteries/1.3V NiMH accumulator Electrode Meter Kit with pH electrode Each for $1,977.74

01-910-520

 
mettler toledo™
ME51302583
pH/Ion Meter, InLab 413 SG electrode (1.8m cable), rubber holster, carry case, four AA 1.5V batteries/1.3V NiMH accumulator Field Kit with pH electrode, rubber holster, carry case Each for $2,113.19
Description & Specifications

Specifications

Accuracy (pH) ±0.01
Accuracy (Temperature) ±0.2°C
Applications Measures ion concentration, pH, mV, temp
Calibration Points (pH) 5
Data Points 200
Dimensions (L x W x H) 1.75 x 3.5 x 8.62 in. (4.5 x 8.89 x 21.9cm)
Range (ISE) 0 to 9999ppm
Range (pH) -2 to 20
Resolution (ISE) 3/4 Digit
Resolution (pH) 0.1/0.01/0.001
Display Type LCD
Inputs BNC (IP67), Cinch (IP67), NTC 30kilohm
Outputs IR to printer or PC via RS232 or USB
Range (mV) 0 to 1999mV
Resolution (mV) 0.1mV
Resolution (Temperature) 0.1°C
Weight (English) 0.71 lb.
Weight (Metric) 325g

  • Portable meter measures ion concentration, pH, mV and temperature
  • Designed for easy, comfortable right- or left-handed operation
  • Choice of three packaging configurations
  • Waterproof and dustproof to IP-67 standard
  • Large backlit display of measured value and settings
  • Rotatable electrode clip that mounts left or right for single-handed operation

  • IP-67 pH/Ion meter with expanded range for the highest demands
  • Continuous indication of electrode and battery status
  • Automatic endpoint and buffer recognition
  • Automatic temperature compensation
  • Three-point calibration with predefined or user-defined buffers
  • Segmented or linear calibration as desired
  • Five-point ion calibration with choice of 6 predefined and one user-defined buffer set
  • Record date, time, time interval
  • Intuitive menu operation using softkeys
  • GLP memory for 200 datapoints
  • Contact-free infrared communication with notebook, printer or PC
  • ON/OFF backlit display

Packaging Options
All packages include wrist strap, four AA batteries, operating instructions, test certificate, and declaration of conformity:

  • Meter Only, without electrode
  • Meter with InLab™ 413-SG electrode and 1.8m cable