Please login to your online account to display your discounted pricing

Mettler Toledo™ SG2 SevenGo™ pH Portable Meter

Built-in data memory and automatic functions for outstanding performance

$1,257.44 - $1,257.44

Specifications

Accuracy (mV) ±1mV
Accuracy (pH) ±0.01
Accuracy (Temperature) ±0.5°C
Battery Type Four AA
Calibration Points (pH) 3
View More Specs
Products
Catalog Number Mfr. No. Type Price Quantity    

01-910-515

 
mettler toledo™
ME51302522
pH Meter with Electrode, 1.2m Cable Each for $1,257.44
Description & Specifications

Specifications

Accuracy (mV) ±1mV
Accuracy (pH) ±0.01
Accuracy (Temperature) ±0.5°C
Battery Type Four AA
Calibration Points (pH) 3
Data Points 30
Display Type LCD
IP Rating 67
Range (mV) 0 to 1999mV
Range (pH) 0 to 14
Resolution (pH) 0.01
For Use With (Application) pH
Inputs BNC
Probe Type NTC
Resolution (mV) 1mV
Resolution (Temperature) 0.1°C
Temperature (English) Operating 32° to 104°F
Temperature (Metric) Operating 0 to 40°C
Weight (English) 11.5 oz.
Weight (Metric) 0.32kg

  • Portable meter measures pH, mV and temperature
  • Designed for easy, comfortable right- or left-handed operation
  • Choice of three packaging configurations
  • Waterproof and dustproof to IP-67 standard
  • Large display of measured value and settings
  • Rotatable electrode clip mounts left or right for single-handed operation

SevenGo SG2

  • IP-67 pH meter with 0 to 14 pH range for routine use
  • Continuous indication of electrode and battery status
  • Automatic endpoint and buffer recognition
  • Automatic temperature compensation
  • Data memory for 30 GLP data points
  • 3-point calibration with predefined or user-defined buffers

Packaging Options
All packages include wrist strap, four AA batteries, operating instructions, test certificate, and declaration of conformity:

  • Meter Only, without electrode
  • Meter with InLab™ 413 SG electrode and 1.2m cable
  • Meter with InLab 413 SG electrode, 1.2m cable, and field case with electrode arm and cover