Mettler Toledo™ SG2 SevenGo™ pH Portable Meter
Built-in data memory and automatic functions for outstanding performance
Manufacturer:  Mettler Toledo™ 51302522
 View more versions of this product
Catalog No. 01-910-515
Specifications
Specifications
32° to 104°F
0 to 40°C
pH Meter with Electrode, 1.2m Cable
3.5 in.
90cm
±0.5°C
3
LCD
pH
0 to 14
IP-67
1.75 in.
NTC
1mV
0.1°C
0.32kg
11.5 oz.
±0.01
±1mV
4 x AA
30
220cm
0.01
Wrist strap, four AA batteries, operating instructions, test certificate, declaration of conformity
BNC
45cm
1.75 x 3.5 in. (45 x 90cm)
0 to 1999mV
Documents