Please login to your online account to display your discounted pricing

Mettler Toledo™ SevenGo™ Series Portable Meter: Model SG78

Characterized by user-friendliness and excellent ergonomics. Mettler Toledo SevenGo Series SG78 Portable Meter is watertight and robust instrument with high measurement accuracy.

$2,460.26 - $3,450.30

Specifications

Accuracy (Conductivity) ±0.5%
Accuracy (mV) ±0.1mV
Accuracy (pH) ±0.002
Accuracy (TDS) ±0.5%
Accuracy (Temperature) ±0.2°C
View More Specs
Products
Catalog Number Mfr. No. Includes Price Quantity    

01-917-137

 
mettler toledo™
ME51302621
Meter, batteries, wrist strap, test certificate, declaration of conformity, InLab Expert-Pro-ISM-IP67 and InLab 738-ISM (both IP67 and 1.8m fixed cable) Each for $3,106.10

01-917-138

 
mettler toledo™
ME51302622
Meter, batteries, wrist strap, test certificate, declaration of conformity, InLab Expert-Pro-ISM-IP67 and InLab 738-ISM (both IP67 and 1.8m fixed cable), field compact case, Rubber holster, 2 SevenGo clips, buffer sachets pH (2 each: 4, 7), conductivity standard sachets (2 each, 1413uS/cm, 12.88mS/cm) and calibration bottles Each for $3,450.30

01-917-136

 
mettler toledo
ME51302620
Meter (no electrodes), batteries, wrist strap, test certificate, declaration of conformity Each for $2,460.26
Description & Specifications

Specifications

Accuracy (Conductivity) ±0.5%
Accuracy (mV) ±0.1mV
Accuracy (pH) ±0.002
Accuracy (TDS) ±0.5%
Accuracy (Temperature) ±0.2°C
Battery Type Four AA
Calibration Points (pH) 5 Points, 7 pre-defined buffer group
Certifications/Compliance GLP/IP 67
Display Type Backlit LCD
IP Rating 67
Outputs IR to printer or PC via RS232 or USB
Range (mV) -1999 to 1999mV
Range (pH) -2 to 19.999
Resolution (pH) 0.1/0.01/0.001
For Use With (Application) Determination of pH, mV, rel. mV or ion concentrations and conductivity, TDS (total dissolved solids), salinity or specific resistance
Inputs pH: BNC/RCA, conductor: LTW 7-pin
Probe Type NTC
Range (Conductivity) 0.01μS/cm to 1000mS/cm
Range (Resistivity) 0 to 100megohm/cm
Range (Salinity) 0 to 80ppt
Range (TDS) 0.01mg/L to 300g/L
Resolution (Conductivity) 0.01/1
Resolution (mV) 0.1mV
Resolution (TDS) 0.01/1
Resolution (Temperature) 0.1°C

  • Easy and intuitive operation
  • pH range: -2 through +19.99
  • Large, easy-to-read display screen
  • Comprehensive range of practical accessories
  • Modern GLP data management
  • Revolutionary Intelligent Sensor Management (ISM™)