Please login to your online account to display your discounted pricing

Mettler Toledo™ SevenGo Duo pro™ pH/Ion/Dissolved Oxygen Meters

Portable dual-channel instruments for the determination of pH, mV, rel.mV, ion concentration and dissolved oxygen.

$2,791.20 - $3,247.54

Specifications

Accuracy (mV) ±0.1mV
Accuracy (pH) ±0.002
Accuracy (Temperature) ±0.2°C
Range (mV) -1999 to 1999mV
Range (pH) -2 to 19.999
View More Specs
Products
Catalog Number Mfr. No. Includes Price Quantity    

01-917-133

 
mettler toledo™
ME51302610
Meter only Each for $2,791.20

01-917-134

 
mettler toledo™
ME51302611
InLab Expert Pro-ISM and InLab 605-ISM and 2-electrode clip Each for $3,247.54
Description & Specifications

Specifications

Accuracy (mV) ±0.1mV
Accuracy (pH) ±0.002
Accuracy (Temperature) ±0.2°C
Range (mV) -1999 to 1999mV
Range (pH) -2 to 19.999
Resolution (pH) 0.1/0.01/0.001
Resolution (Temperature) 0.1°C

 

  • Five-point pH calibration with a choice of seven predefined and one uer-sefined buffer set
  • Data storage for 500 GLP data records with time/date, sensor ID and SN, and user and sample IDs
  • Contact-free IR communication
  • Intelligent Sensor Managment (ISM™)