Please login to your online account to display your discounted pricing

Mettler Toledo™ SevenGo Duo pro™ Lab Meters

Professional dual-channel instrument for special analyses, particularly in environmental, chemical and food and beverage industries. Mettler Toledo SevenGo Duo pro Lab Meters are portable meters for parallel determination of pH, mV, rel. mV or ion concentrations, and dissolved oxygen (DO).

$2,105.18 - $3,312.80

Specifications

Accuracy (mV) ±0.1mV
Accuracy (pH) ±0.002
Accuracy (Temperature) ±0.2°C
Battery Type Four AA
Calibration Points (pH) 5
View More Specs
Products
Catalog Number Mfr. No. Includes Type Price Quantity    

01-915-700

 
mettler toledo™
ME51302661
Lab Meter Meter Only Each for $2,105.18

01-915-703

 
mettler toledo™
ME51302664
Lab Meter, InLab Expert Pro-ISM and InLab OptiOx (both IP67, 5m cable), OptiOx protective guard, Rubber holster, calibration sachets, sample bottles and field compact case Meter Field Kit with Electrodes and Case Each for $3,312.80

01-915-702

 
mettler toledo™
ME51302663
Lab Meter, InLab Expert Pro-ISM and InLab OptiOx (both IP67, 1.8m cable), OptiOx protective guard, Rubber holster, calibration sachets, sample bottles and field compact case Meter Field Kit with Electrodes and Case Each for $3,176.82

01-915-701

 
mettler toledo™
ME51302662
Lab Meter, Expert Pro-ISM and InLab OptiOx (both IP67, 1.8m cable) Meter Electrode Kit Each for $3,161.47
Description & Specifications

Specifications

Accuracy (mV) ±0.1mV
Accuracy (pH) ±0.002
Accuracy (Temperature) ±0.2°C
Battery Type Four AA
Calibration Points (pH) 5
Dimensions (L x W x H) 8.66 x 3.54 x 1.77 in. (22 x 9 x 4.5cm)
IP Rating 67
Outputs IR to printer or PC via RS232 or USB
Range (mV) -1999 to 1999mV
Range (pH) -2 to 19.999
Resolution (pH) 0.001
Inputs BNC (>1012Ohm), RCA (cinch), (both IP67)
Resolution (mV) 0.1mV
Resolution (Temperature) 0.1°C
Temperature (English) Operating 32° to 104°F
Temperature (Metric) Operating 0 to 40°C

Featuring

  • Either linear or segmented 5-point pH calibration, with selection from seven predefined and one user-defined buffer groups
  • Dissolved oxygen mode based on optical Rugged Dissolved Oxygen (RDO™) technology
  • Manual or automatic atmospheric pressure compensation using built-in barometer
  • Modern data management for 500 GLP data records with date/time, sensor ID and serial number, and user and sample ID
  • Contactless infrared interface for data transmission to a printer or PC
  • Intuitive, user-friendly operation using menu guidance in 10 languages and backlit display
  • Rugged watertight housing (IP67)
  • Intelligent sensor management system (ISM™) for automatic access to up-to-date calibration data

 InLab™ OptiOx for Dissolved Oxygen

  • Optical oxygen sensor based on RDO (Rugged Dissolved Oxygen) technology
  • Easy-to-replace OptiOx sensor caps
  • Robust and manageable design
InLab Expert Pro-ISM for pH
  • Low-maintenance XEROLYT™ polymer electrolyte with two open reference connections
  • Robust, chemically resistant PEEK shaft
  • Long-lasting ARGENTHAL™ reference system