missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
METTLER TOLEDO™ SevenExcellence™ S400 Benchtop pH Meter
The S400 meter measures pH, redox/ORP, and ISFET. It can be expanded with additional modules to measure conductivity, resistivity, DO, BOD, ISE, salinity, and TDS. It has an intuitive touchscreen, the ability to create measurement shortcuts, and can export data to a printer, Excel, or LIMS system.
$4055.48 - $5056.48
Specifications
Accuracy (mV) | ±0.1 |
---|---|
Accuracy (pH) | ±0.002 |
Accuracy (Temperature) | ±0.1°C |
Outputs | USB, RS-232, Ethernet |
Range (pH) | -2 to 20 |
Catalog Number | Mfr. No. | Includes | Model | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Model | Price | Quantity | ||||
01-910-901
|
METTLER TOLEDO™
30046241 |
S400 pH meter; InLab Expert Pro-ISM electrode; electrode holder; 2 buffer sachets for pH 4.01, 7, 9.21 and 10.01; EasyDirect pH activation key (pH data management software); test certificate; pH measurement guide | S400 Kit - Standard Applications |
Each for $4,713.28
|
|
||||
01-910-900
|
METTLER TOLEDO™
30046240 |
S400 pH meter; electrode holder; 2 buffer sachets for pH 4.01, 7, 9.21 and 10.01; EasyDirect pH activation key (pH data management software); test certificate | S400 pH Meter |
Each for $4,055.48
|
|
||||
01-910-902
|
METTLER TOLEDO™
30046242 |
S400 pH meter; InLab Routine Pro-ISM electrode; electrode holder; ; 2 buffer sachets for pH 4.01, 7, 9.21 and 10.01; EasyDirect pH activation key (pH data management software); test certificate; pH measurement guide; Multipin-BNC/RCA electrode cable 1.2 m | S400 Kit - Bio & Life Science Applications |
Each for $5,056.48
|
|
||||
01-910-921
|
METTLER TOLEDO™
30092983 |
S400 pH meter; InLab Expert Pro-ISM electrode; electrode holder; uMix automatic magnetic stirrer; 2 buffer sachets for pH 4.01, 7, 9.21 and 10.01; EasyDirect pH activation key (pH data management software); test certificate; pH measurement guide | S400 Kit - Standard Applications + Automatic Stirring |
Each for $4,244.23
|
|
||||
01-910-920
|
METTLER TOLEDO™
30092982 |
S400 pH meter; InLab Ultra-Micro-ISM electrode; electrode holder; 2 buffer sachets for pH 4.01, 7, 9.21 and 10.01; EasyDirect pH activation key (pH data management software); test certificate; pH measurement guide; Multipin-BNC/RCA electrode cable 1.2 m | S400 Kit - Small Samples |
Each for $4,381.52
|
|
||||
Description
- pH, Redox/ORP, and ISFET meter than can be expanded with additional measurement modules to measure conductivity, resistivity, dissolved oxygen, ISE, salinity, TDS, and BOD
- Easily create measurement and calibration shortcuts
- 7 inch color touch screen with intuitive menu navigation
- Compatible with a wide range of electrodes for any application
- *Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
- Meter connects to electrodes via BNC; S7 to BNC cables available
- Export data to a printer, Excel file, or LIMS system
- Automation available for measurements and cleaning in high-throughput applications
- Manage user permissions and access to ensure data integrity
- Compatible with LabX software for advanced data management and 21CFR part 11 regulatory compliance
- Optional comprehensive qualification package available including IQ/OQ/PQ
- Preventative maintenance and service available through METTLER TOLEDO Service
Specifications
±0.1 | |
±0.1°C | |
-2 to 20 | |
Max. 5 points, 8 pre-defined and 20 user-defined buffer group | |
0.1/1mV | |
0.1°C | |
20000 | |
0.001 to 1 |
±0.002 | |
USB, RS-232, Ethernet | |
0.001, 0.01, 0.1 | |
-2000 to 2000 | |
-30°C to +130°C | |
TFT | |
0.001μS/cm to 2000mS/cm | |
13 pre-defined and 20 user-defined standard |