missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
METTLER TOLEDO™ SevenExcellence™ S400 pH Benchtop Meter
The S400 meter measures pH, redox/ORP, and ISFET. It can be expanded with additional modules to measure conductivity, resistivity, DO, BOD, ISE, salinity, and TDS. It has an intuitive touchscreen, the ability to create measurement shortcuts, and can export data to a printer, Excel, or LIMS system.
Manufacturer: METTLER TOLEDO™ 30046240
Description
- pH, Redox/ORP, and ISFET meter than can be expanded with additional measurement modules to measure conductivity, resistivity, dissolved oxygen, ISE, salinity, TDS, and BOD
- Easily create measurement and calibration shortcuts
- 7 inch color touch screen with intuitive menu navigation
- Compatible with a wide range of electrodes for any application
- Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
- Meter connects to electrodes via BNC; S7 to BNC cables available
- Export data to a printer, Excel file, or LIMS system
- Automation available for measurements and cleaning in high-throughput applications
- Automation available for measurements and cleaning in high-throughput applications
- Manage user permissions and access to ensure data integrity
- Compatible with LabX software for advanced data management and 21CFR part 11 regulatory compliance
- Optional comprehensive qualification package available including IQ/OQ/PQ
Specifications
20000 | |
USB, RS232 and Ethernet | |
13 pre-defined and 20 user-defined standard | |
Single-channel | |
Chinese, Chinese (TW), Croatian, Czech, Danish, Dutch, English (GB), English (US), French, German, Hungarian, Indonesian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese (BR), Russian, Slovak, Slovenian, Spanish, Swedish, Thai, Turkish, Vietnamese | |
S400 pH Meter | |
-2000 to 2000 mV | |
0.001, 0.01, 0.1 | |
0.1/1mV |
TFT | |
-30°C to +130°C | |
Max. 5 points, 8 pre-defined and 20 user-defined buffer group | |
S400 pH meter; electrode holder; 2 buffer sachets for pH 4.01, 7, 9.21 and 10.01; EasyDirect pH activation key (pH data management software); test certificate | |
250 analyses (20.000 data points) | |
0.001μS/cm to 2000mS/cm | |
-2 to 20 | |
0.001 to 1 | |
0.1°C |
Promotions
-
Earn Up to $2,000 in Accessories with Purchase
Buy METTLER TOLEDO balances or analytical instruments, you’ll get up to $2,000 to spend on accessories.: 06/30/21
-
Earn Up to 5X the Fisher Scientific Buck$ with Purchase
Bundle your purchase of qualifying METTLER TOLEDO balances or analytical instruments to earn more.: 06/30/21