Mettler Toledo™ SevenExcellence™ S400 pH/mV Meters
Intuitive touchscreen operation and high measurement performance combined with a complete security package for safe results and solid compliance. Mettler Toledo SevenExcellence S400 pH/mV Meter is a single-channel instrument which is modularly expandable with additional measurement parameters at any time — conductivity, ion or dissolved oxygen.
Manufacturer:  Mettler Toledo™ 30046243
Catalog No. 01-910-903
Description
Description
  • Professional calibration support, including automatic calibration with optional Rondolino sample changer
  • Large 7 in. color display and intuitive menu guidance in 10 languages
  • Flexible method concept for high reproducibility and security
  • Extra security thanks to sophisticated user management and Intelligent Sensor Management (ISM)
  • Integrated interfaces (USB, RS-232 and Ethernet) for data exchange
  • Optional comprehensive qualification package including IQ/OQ/PQ
Specifications
Specifications
±0.1
±0.1°C
Max. 5 points, 8 pre-defined and 20 user-defined buffer group
TFT
S400-ISFET
0.001μS/cm to 2000mS/cm
-2 to 20
0.001 to 1
0.1°C
±0.002
13 pre-defined and 20 user-defined standard
20000
S-400 Meter; InLab 490; guide to pH measurements; 2 buffer sachets for pH 4.01, 7, 9.21 and 10
USB, RS232 and Ethernet
-2000 to 2000 mV
0.001, 0.01, 0.1
0.1/1mV
-30°C to +130°C
Documents