Please login to your online account to display your discounted pricing

Mettler Toledo™ Seven2Go™ Pro pH/mV and Ion Meter: S8

Ideal for mobile applications in the laboratory, at-line, and outdoors

$1,697.20 - $2,404.38

Specifications

Range (pH) -2 to 20
Accuracy (pH) ±0.002
Resolution (pH) 0.001/0.01/0.1
Accuracy (Ion) ±0.5%
Range (mV) -2000 to +2000mV
View More Specs
Products
Catalog Number Mfr. No. Type Price Quantity    

01-913-981

 
mettler toledo™
ME30207874
Seven2Go Pro S8 pH/mV and Ion Meter Each for $1,697.20

01-913-982

 
mettler toledo™
ME30207875
Seven2Go Pro S8 pH/mV and Ion Meter: Standard Kit Each for $2,018.15

01-913-983

 
mettler toledo™
ME30207877
Seven2Go Pro S8 pH/mV and Ion Meter: Field Kit Each for $2,197.66

01-913-984

 
mettler toledo™
ME30207878
Seven2Go Pro S8 pH/mV and Ion Meter: Biotech Kit Each for $1,958.31

01-913-985

 
mettler toledo™
ME30207879
Seven2Go Pro S8 pH/mV and Ion Meter: Fluoride Kit Each for $2,404.38
Description & Specifications

Specifications

Range (pH) -2 to 20
Accuracy (pH) ±0.002
Resolution (pH) 0.001/0.01/0.1
Accuracy (Ion) ±0.5%
Range (mV) -2000 to +2000mV
Accuracy (mV) ±0.1
Resolution (mV) 0.1/1
Temperature (Metric) Range -5° to +130°C (ATC); -30° to +130°C (MTC)
Accuracy (Temperature) ±0.2°C if T: 0° to 105°C; ±0.5°C if T<0°C or T>105°C
Resolution (Temperature) 0.1°C
Calibration Points (pH) 5-Point
Certifications/Compliance ISO 9001, ISO 14001
Display Type LCD
IP Rating IP67
Inputs BNC (1012 Ohm)/RCA (cinch)/2mm banana
Data Management Details Micro-USB (USB Device)
Data Points 2000 (GLP conform)
Battery Type Four 1.5V AA, 1.3V NiMH acc.
Battery Life 200 to 250 hours
Dimensions (L x W x H) 222 x 70 x 35mm
Weight (Metric) 290g

The Seven2Go portable meters are simple, reliable and are engineered to provide users with fast quality data, one-handed operation and durability.

  • New intuitive menu guarantees out-of-the-box operation for anyone
  • T-Pad provides comfort and improves the speed with which you move through menus
  • Smart button arrangement for faster navigation
  • Automatically recognizes InLab ISM™ (Intelligent Sensor Management) sensor and stores sensor ID, serial number, and last calibration data, thereby reducing chances of error
  • IP67 protection and drop test resistance make these meters weatherproof (withstand harsh and demanding environments)
  • Supports 12 languages and offers a user interface that uses complete words and sentences
  • Lack of cryptic expressions simplify operation and reduces strain on the user
  • Sunlight-proof: direct sunlight enhances screen's contrast and guarantees an excellent readability even under bright and sunny conditions
  • StatusLight™, an easy to understand traffic light helps to check if your meter is ready or not
  • 5-Point calibration (pH and Ion)
  • Linear and segmented calibration methods (pH and Ion)
  • Graphic LCD display
  • Ability to perform automatic, manual and time-dependent endpoint recording
  • Perform serial measurements in user-defined time interval
  • Acoustic and visual endpoint signal
  • Other features include: Time and date option, three user levels (Routine, Expert, Outdoor), Calibration reminder, Sensor ID, SN, Sample ID, User ID, User-defined alarm limits, PIN protection
  • Includes eight predefined buffer groups and one user-defined Ion-standards
  • User-defined buffer group
  • Automatic buffer recognition
  • Automatic/Manual temperature capture
Specifications:
  • pH Measuring Range: -2 to 20
  • pH Resolution: 0.001/0.01/0.1
  • pH Accuracy: ±0.002
  • ORP/Redox Measuring Range: -2000 to +2000mV
  • ORP/Redox Resolution: 0.1/1
  • ORP/Redox Accuracy: ±0.1
  • Units: mV, rel. mV
  • Ion Measuring range: 1.00e-09 to 9.99e+09
  • Ion Resolution: 3 or 4 digits (auto-range)
  • Ion Accuracy: ±0.5%
  • Units: mg/L, mmol/L, mol/L, %, ppm, pX
  • Temperature Measuring Range: -5° to +130°C (ATC); -30° to +130°C (MTC)
  • Temperature Resolution: 0.1°C
  • Temperature Accuracy: ±0.2°C if T: 0° to 105°C; ±0.5°C if T<0°C or T>105°C
  • Memory Size: 2000 (GLP conform)
  • Sensor Inputs: BNC (1012 Ohm)/RCA (cinch)/2mm banana
  • Communication interfaces: Micro-USB (USB Device)
  • Power Supply: Four 1.5 V AA, 1.3 V NiMH acc. or USB
  • Operating Hours: 200 to 250 hours
  • Dimensions: 222 x 70 x 35mm
  • Weight: 290g
Applications:
  • S8-Standard kit: Wide range of samples, also suited for emulsions and suspensions.
  • S8-Field kit: Wide range of samples, also suited for emulsions and suspensions; ideal for measurements on the go.
  • S8-Biotech kit: Fast results in wide range of samples including harsh chemicals.
  • S8-Fluoride kit: Aqueous samples, tooth paste and mouth wash.
Kits Available:


  • Cat. No. 01-913-981, S8-Meter: Instrument, LabX direct pH, USB cable, electrode clip, base accessory, operating instructions, quick guide, test report, declaration of conformity and batteries.
  • Cat. No. 01-913-982, S8-Standard kit: Instrument, LabX direct pH, USB cable, electrode clip, InLab™ Expert Pro-ISM-IP67, buffer sachets (for pH 4.01, 7.00,9.21 and 10.00), base accessory, operating instructions, quick guide, test report, declaration of conformity and batteries.
  • Cat. No. 01-913-983, S8-Field kit: Instrument, LabX direct pH, USB cable, electrode clip, InLab™ Expert Pro-ISM-IP67, buffer sachets (for pH 4.01, 7.00,9.21 and 10.00), bottles for calibration, uGo™ carrying case, base accessory, operating instructions, quick guide, test report, declaration of conformity and batteries.
  • Cat. No. 01-913-984, S8-Biotech kit: Instrument, LabX direct pH, USB cable, electrode clip, InLab™ Routine Pro ISM (includes cable), buffer sachets (for pH 4.01, 7.00,9.21 and 10.00), bottles for calibration, uGo™ carrying case, base accessory, operating instructions, quick guide, test report, declaration of conformity and batteries.
  • Cat. No. 01-913-985, S8-Fluoride kit: Instrument, LabX direct pH, USB cable, electrode clip, perfectION Fluoride, base accessory, operating instructions, quick guide, test report, declaration of conformity and batteries.

ISO 9001, ISO 14001