missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mettler Toledo™ Seven2Go™ pH/mV Meter: S2

Ideal for mobile applications in the laboratory, at-line, and outdoors

$875.80 - $1,517.69

Specifications

Range (pH) -2 to 20
Accuracy (pH) ±0.01
Resolution (pH) 0.01 to 0.1
Range (mV) -1999 to 1999 mV
Accuracy (mV) ±1
View More Specs

Products 5
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
01-913-965
View Documents
Mettler Toledo™
30207949
Seven2Go S2 pH/mV Meter
Each for $875.80
 
01-913-966
View Documents
Mettler Toledo™
30207950
Seven2Go S2 pH/mV Meter: Standard Kit
Each for $1,343.62
 
01-913-967
View Documents
Mettler Toledo™
30207951
Seven2Go S2 pH/mV Meter: Field Kit w/ pH probe
Each for $1,392.58
 
01-913-968
View Documents
Mettler Toledo™
30207952
Seven2Go S2 pH/mV Meter: Food Kit
Each for $1,517.69
 
01-913-969
View Documents
Mettler Toledo™
30207953
Seven2Go S2 pH/mV Meter: Light Kit
Each for $979.16
 
Description

Description

The Seven2Go portable meters are simple, reliable and are engineered to provide users with fast quality data, one-handed operation and durability.
  • New intuitive menu guarantees out-of-the-box operation for anyone
  • T-Pad provides comfort and improves the speed with which you move through menus
  • Smart button arrangement for faster navigation
  • Automatically recognizes InLab ISM™ (Intelligent Sensor Management) sensor and stores sensor ID, serial number, and last calibration data, thereby reducing chances of error
  • IP67 protection and drop test resistance make these meters weatherproof (withstand harsh and demanding environments)
  • 5-Point calibration (pH)
  • Segmented LCD display
  • Four predefined buffer groups
  • Automatic buffer recognition
  • Linear and segmented calibration methods
  • With user-defined buffer group
  • Ability to perform manual and time-dependent endpoint recording
  • Perform serial measurements in user-defined time interval
  • Visual endpoint signal
  • Automatic/Manual temperature capture
Specifications:
  • pH Measuring Range: -2 to 20
  • pH Resolution: 0.01/0.1
  • pH Accuracy: ±0.01
  • ORP/Redox Measuring Range: -1999 to +1999mV
  • ORP/Redox Resolution: 1
  • ORP/Redox Accuracy: ±1
  • Units: mV, rel. mV
  • Temperature Measuring Range: -5° to +105°C
  • Temperature Resolution: 0.1°C
  • Temperature Accuracy: ±0.5°C
  • ISM: Light version
  • Memory Size: 200
  • Sensor Inputs: BNC (1012 Ohm)/RCA (cinch)
  • Power Supply: Four 1.5 V AA or 1.3 V NiMH acc.
  • Operating Hours: 250 to 400 hours
  • Dimensions: 222 x 70 x 35mm
  • Weight: 270g
Applications:
  • S2-Standard kit: Wide range of samples including emulsions and suspensions.
  • S2-Field kit: Wide range of samples including emulsions and suspensions; ideal for measurements on the go.
  • S2-Food kit: Solid and semi-solid samples like cheese and meat.
  • S2-Light kit: Academic and educational purposes.
Kits Available:
  • Cat. No. 01-913-965, S2-Meter: Instrument, electrode clip, operating instructions, quick guide, test report, declaration of conformity and batteries.
  • Cat. No. 01-913-966, S2-Standard kit: Instrument, electrode clip, InLab™ Expert Pro-ISM-IP67, buffer sachets (pH 4.01, 7.00, 9.21 and 10.00), operating instructions, quick guide, test report, declaration of conformity and batteries.
  • Cat. No. 01-913-967, S2-Field kit: Instrument, electrode clip, InLab™ Expert Pro-ISM-IP67, buffer sachets (pH 4.01, 7.00, 9.21 and 10.00), bottles for calibration, uGo™ carrying case, operating instructions, quick guide, test report, declaration of conformity and batteries.
  • Cat. No. 01-913-968, S2-Food kit: Instrument, electrode clip, InLab™ Solids Pro-IP67, buffer sachets (pH 4.01, 7.00, 9.21 and 10.00), bottles for calibration, uGo™ carrying case, operating instructions, quick guide, test report, declaration of conformity and batteries.
  • Cat. No. 01-913-969, S2-Light kit: Instrument, electrode clip, InLab™ Versatile Pro, buffer sachets (pH 4.01, 7.00, 9.21 and 10.00), operating instructions, quick guide, test report, declaration of conformity and batteries.

Certifications

ISO 9001, ISO 14001

Specifications

Specifications

-2 to 20
0.01 to 0.1
±1
-5°C to +105°C
0.1°C
ISO 9001, ISO 14001
IP-67
200 (Memory)
250 to 400 hr.
±0.01
-1999 to 1999 mV
1
±0.5°C
5
LCD
BNC (1012 Ohm)/RCA (cinch) (sensor)
4 x AA or 1.3V NiMH acc.
222 x 70 x 35 mm
SDS