missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mettler Toledo™ pH Electrodes

For pH measurements.

$615.00 - $1,180.00

Specifications

For Use With (Equipment) pH meter
Type pH Electrode
View More Specs

Products 6
Catalog Number Mfr. No. Membrane Filling Solution Type Model pH Range Price Quantity  
Catalog Number Mfr. No. Membrane Filling Solution Type Model pH Range Price Quantity  
01920150
View Documents
Mettler Toledo™
52000497
LR3 Glass Polymer InLab 1010 0 to 14
Each for $615.00
 
01920156
View Documents
Mettler Toledo™
59902917
LoT Glass DPA gel InLab™ Flow 1 to 11
Each for $675.00
 
01920158
View Documents
Mettler Toledo™
59902993
LoT Glass 3mol/L KCl AgCl saturated InLab LoT403-M8-S7/120 1 to 11
Each for $635.00
 
01920167
View Documents
Mettler Toledo™
59904572
U Glass 3mol/L KCl AgCl saturated InLab™ NMR 0 to 14
Each for $1,180.00
 
01920144
View Documents
Mettler Toledo™
51340262
HA Glass 3mol/L KCl HA405-60-M8-S7/400 0 to 14
Each for $850.00
 
01920169
View Documents
Mettler Toledo™
59904764
HA Glass 3mol/L KCl HA425-60-S7/120 0 to 14
Each for $1,095.00
 
Description

Description

  • BNC connectors of 1.0m
  • Red S7 connectors compatible with gray S7 connectors

Order Info

Metal Half-Cells (01-920-162, -163 and -164) and U-Electrodes (09-320-200 and -201) also available separately.

Specifications

Specifications

pH meter
pH Electrode
SDS