Mettler Toledo™ NewClassic ML Precision Balances
NewClassic ML Balances With External Adjustment
Manufacturer:  Mettler Toledo™ ME11145255
 View more versions of this product
Catalog No. 01-910-224
Specifications Description & Specifications
Specifications
220g
29 x 18.4 x 18.7cm
7.3 in.
Keys
ABS Plastic, Die-cast Aluminum, Stainless Steel
29cm
10w
10 to 30°C
1.5 sec.
6.17 lb.
2.8kg
7.2 in.
50 to 86°F
0.3A
12cm
Backlit LCD
1 Year
11.4 in.
±0.002g
ML203E
18.7cm
ISO 9001/17025, UL, CSA, OIML, CE
Draft shield, protective cover, universal AC adapter, declaration of conformity, quick guide, operating instructions
RS232
0.001g
0.001g
3g
18.4cm
100/240V
Precision balance