Please login to your online account to display your discounted pricing
Mettler Toledo™ NewClassic ML Precision Balances
NewClassic ML Balances With External Adjustment
$2,095.34 - $3,050.81
Specifications
Material ABS Plastic, Die-cast Aluminum, Stainless Steel
Connection Type RS232
Display Type Backlit LCD
Certifications/Compliance ISO 9001/17025, UL, CSA, OIML, CE
View More Specs
Products
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity    
01-910-224
mettler toledo™
ME11145255
ML203E 1.5 sec. 220g 0.001g Each for $2,095.34
01-910-229
mettler toledo™
ME11145333
ML400E 1 sec. 4200g 0.01g Each for $3,050.81
01-910-228
  Accessories
mettler toledo™
ME11145327
ML300E 1 sec. 3200g 0.01g Each for $2,271.34
Description & Specifications
Specifications
Material ABS Plastic, Die-cast Aluminum, Stainless Steel
Connection Type RS232
Display Type Backlit LCD
Certifications/Compliance ISO 9001/17025, UL, CSA, OIML, CE
  • MonoBloc™ proven weighing technology for fast and accurate results
  • Full independence from power supply for 8 hours
  • Protects the weighing cell against excess weight overload
  • RS232 interface for connection to PC or printer. No additional software needed
  • Easy running of balance diagnostics test e.g. keypad test, repeatability test
  • OIML Approved versions of all models available
  • Adjustment at a keystroke with an internal weight
  • Protective cover encloses the entire housing protecting the balances from stains and scratches
Piece Counting, Percent Weighing, Check Weighing, Dynamic Weighing, Statistics
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.