Please login to your online account to display your discounted pricing
Mettler Toledo™ NewClassic ML Analytical Balances with Draft Shield
The mobile all-rounder for easy operation and small budgets
$3,675.22 - $3,830.27
Specifications
Material Solid Metal
Connection Type RS232
Display Type Backlit LCD
Certifications/Compliance ISO 9001, UL, CSA
View More Specs
Products
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity    
01-910-221
mettler toledo™
ME11145210
ML54 2 sec. 52g 0.1mg Each for $3,675.22
01-910-222
mettler toledo™
ME11145216
ML104 2 sec. 120g 0.1mg Each for $3,830.27
Description & Specifications
Specifications
Material Solid Metal
Connection Type RS232
Display Type Backlit LCD
Certifications/Compliance ISO 9001, UL, CSA
NewClassic ML balances have everything a basic balance needs: an ergonomic design with a brilliant display, easy-to-clean glass draft shield, an intuitive user interface, multiple built-in applications, battery operation for full mobility and good weighing performance and robustness.
  • MonoBloc: proven weighing technology for fast and accurate results
  • Battery operation: full independence from power supply for 8 hours
  • Adjustment at a keystroke with external weight and internal weight
  • SmartTrac shows how much of the entire weighing range has been used
  • Protection of main balance configurations against manipulation
  • Overload protection, protects the weighing cell against excess weight overload
  • Diagnostics: easy running of balance diagnostics test (e.g., keypad test, repeatability test)
  • Connectivity RS-232 interface for connection to PC or printer
  • Protective cover encloses the entire housing protecting the balances from stains and scratches
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.