Mettler Toledo™ NewClassic ML 0.1g Balances
Ideal for work in field or whenever power supply is hard to find. Mettler Toledo NewClassic ML 0.1g Balances come with brilliant backlit display that can be used even under extreme lighting conditions. Built-in battery slot fits eight standard AA batteries and supports 8 hours of use.
$1,470.93 - $1,470.93
Specifications
Connection Type RS232
Display Type Backlit LCD
Certifications/Compliance UL, CSA
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
01-910-230 Accessories Mettler Toledo™
ME11143274
ML601E 1 sec. 620g 0.1g Each for $1,470.93
Specifications
Description
  • Ergonomic design with multiple built-in applications
  • Direct connection: connects balance to a PC via the RS232 slot
  • Easy-to clean glass draft shield
  • Internal adjustment at a keystroke
  • Protective cover, Overload protection
  • Below weighing hook for weighing below the balance
  • MonoBloc™ weighing technology
  • Built in date and time function
  • Programmable one Smart key for shortcut access to application
  • Protection of main balance configurations
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
RS232
Backlit LCD
UL, CSA