Please login to your online account to display your discounted pricing
Mettler Toledo™ Ion-Selective Combination Electrode Cable Connections
Manufacturer: mettler toledo™  ME52300046
 View more versions of this product
Catalog No. 01-913-246
  • / Each
Description & Specifications
Specifications
Length (English) Cable 47.2 in.
Length (Metric) Cable 1.2m
Connector Types S7 to BNC (IP67)
For Use With MP120, MP125 and MP130 portable meter
Type Cable Connection