Please login to your online account to display your discounted pricing

Mettler Toledo™ InLab™ Versatile Series pH and Pro pH Electrodes

pH/ATC combination electrode. The InLab™ Versatile Series pH and Pro pH Electrode is an excellent alternative to open or movable sleeve junction with a replaceable junction.

$111.81 - $495.37

Specifications

Applications Alternative for measurements in simple samples, offer an outstanding price/performance ratio
Junction Ceramic
View More Specs
Products
Catalog Number Mfr. No. For Use With Type Price Quantity    

01-913-203

 
mettler toledo™
ME51340267
InLab designations versatile and versatile Pro pH electrode Replaceable junction tool Each for $137.45

01-913-204

 
mettler toledo™
ME51340268
InLab designations versatile and versatile Pro pH electrode Set of replaceable junctions Pack of 10 Each for $111.81

01-911-483

 
mettler toledo™
ME51343030
pH meter pH combination electrode Each for $327.33

01-911-484

 
mettler toledo™
ME51343031
pH meter pH/ATC combination electrode Each for $495.37
Description & Specifications

Specifications

Applications Alternative for measurements in simple samples, offer an outstanding price/performance ratio
Junction Ceramic

InLab Versatile

  • 2-in-1 sensor (pH, Reference)
  • Refillable
  • Robust polysulfone shaft
  • BNC connector
InLab Versatile Pro
  • 3-in-1 sensor (pH, Reference, Temperature)
  • Integrated 30kOhm probe
  • Refillable
  • Robust polysulfone shaft
  • BNC and RCA connectors

 

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.