missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
METTLER TOLEDO™ InLab™ Reference Electrodes
A reference electrode to be paired with half-cell measurement electrodes.
Supplier: METTLER TOLEDO™ 51343191
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- For use with half-cell measuring electrodes
Accessories & Care
- S7 connector type: Cable S7 to 2 mm Banana (30281923) required
- InLab Reference: ceramic junction
- InLab Reference Plus: moveable PTFE sleeve junction
- InLab Reference Flow: triple ceramic junction
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
S7 | |
Glass | |
Half-Cell Reference Electrode | |
For routine applications and measurements in Ion-selective half cells | |
InLab™ Reference Plus | |
32°F to 140°F | |
4.72 in. | |
0.47 in. |
PTFE Sleeve | |
0°C to 60°C | |
3mol/L KCl | |
AAg/AgCl | |
Gel | |
0°C to 60°C | |
12 cm | |
1.2 cm |