missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mettler Toledo™ InLab™ Conductivity Sensors
Sensors with Intelligent Sensor Management (ISM) technology. Mettler Toledo InLab Conductivity Sensors with integrated NTC 30kOhm temperature sensor enables automatic temperature compensation.
Manufacturer: Mettler Toledo™ 51344110
Description
- High-performance conductivity cells with fixed cable (LTW)
- Waterproof to IP67, ideal for field measurements
- Easy to maintain
Specifications
0 to 100°C | |
Mini DIN (IP67) | |
12mm | |
4.72 in. | |
120mm | |
Epoxy shaft/4 Graphite poles | |
Dry | |
Conductivity | |
InLab 738-ISM |
0.57cm-1 | |
0.47 in. | |
SevenGo Duo™ SG23 and SevenGo Duo pro™ SG78 Dual Channel Portable Meters | |
70.86 in. | |
1.8m | |
0.01 to 1000μS/cm | |
0° to 100°C | |
NTC 30kilohm |