missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mettler Toledo™ InLab™ Conductivity Probes
Ideal sensor for general conductivity measurements. Mettler Toledo InLab Conductivity Probes have integrated temperature sensors that enable correction to a reference temperature.
Manufacturer: Mettler Toledo™ 51344031
Description
Mettler Toledo offers a wide rand of conductivity cells to meet any laboratory application. Cells are shipped in a ready-to-use condition and include cable and connector.
- For all ranges of conductivity
- Glass or epoxy shaft
- Temperature probe (NTC 30kω)
- Included certificate specifies the nominal and actual cell constant
Specifications
General conductivity measurements | |
0°C to 100°C | |
0.23 in. | |
39.3 in. | |
Glass | |
2m attached cable |
Integrated Temperature Sensor | |
Mini-DIN | |
6 mm | |
1.2 m | |
0.01 to 112 mS/cm | |
InLab 752-6mm |