Mettler Toledo™ FiveEasy Plus™ FEP30 Conductivity Meter
Economical and reliable solution for easy conductivity measurement
Manufacturer:  Mettler Toledo™ ME51302936
 View more versions of this product
Catalog No. 01-911-304
Specifications Description & Specifications
Specifications
FEP30-Kit with LE703 probe
±0.5%
0 to 19.99psu
1
0.1°C
LCD
Mini-DIN
9VDC
0.1μS/cm to 199.9mS/cm
0.1mg/L to 199.9g/L
Conductivity
0° to 100°C
±0.3°C
99
RS232
100/240V