Please login to your online account to display your discounted pricing

FiveEasy Plus™ FEP20 pH Meter


Ideal for anyone who needs an economic and reliable solution for easy pH measurement

Manufacturer: mettler toledo  ME30089749

 View more versions of this product

Catalog No. 01-913-997

  • / Each
 

Description & Specifications

Specifications

Item Description FiveEasy Plus FEP20 pH Meter: Micro Kit
Includes Meter, operating manual, Quick guide, power adapter, LE422 pH electrode, electrode arm, 2X pH 4.01 and 2X pH 7.00 buffer sachets
Range (pH) 0.00 to 14.00
Resolution (pH) 0.01
Accuracy (pH) ±0.01
Range (mV) -1999 to 1999mV
Resolution (mV) 1mV
Accuracy (mV) ±1mV
Range (Temperature) 0° to 100°C
Resolution (Temperature) 0.1°C
Accuracy (Temperature) ±0.5°C
Display Type LCD
Data Management Details Up to 99 data storage
Outputs RS-232
Certifications/Compliance CE marked, ISO 9001, ISO 14001
Dimensions (L x W x H) 200 x 175 x 52mm
Weight (Metric) 0.6kg
Electrical Requirements 100 to 240V, 50 to 60Hz, 9VDC
Height (Metric) 52mm
Length (Metric) 200mm
Material ABS/PC
Temperature Compensation Automatic or Manual
Temperature Range (English) 32° to +212°F
Temperature Range (Metric) 0° to +100°C
Width (Metric) 175mm