Please login to your online account to display your discounted pricing

Mettler Toledo™ FiveEasy Plus™ FEP20 pH Meter

Ideal for anyone who needs an economic and reliable solution for easy pH measurement

$1,060.75 - $1,225.43

Specifications

Range (pH) 0.00 to 14.00
Resolution (pH) 0.01
Accuracy (pH) ±0.01
Range (mV) -1999 to 1999mV
Resolution (mV) 1mV
View More Specs
Products
Catalog Number Mfr. No. Item Description Price Quantity    

01-911-301

 
mettler toledo™
ME51302914
FiveEasy Plus FEP20 Basic pH Meter Each for $1,060.75

01-913-997

 
mettler toledo
ME30089749
FiveEasy Plus FEP20 pH Meter: Micro Kit Each for $1,158.67

01-913-996

 
mettler toledo
ME30089748
FiveEasy Plus FEP20 pH Meter: TRIS Kit Each for $1,225.43
Description & Specifications

Specifications

Range (pH) 0.00 to 14.00
Resolution (pH) 0.01
Accuracy (pH) ±0.01
Range (mV) -1999 to 1999mV
Resolution (mV) 1mV
Accuracy (mV) ±1mV
Temperature Range (Metric) 0° to +100°C
Temperature Range (English) 32° to +212°F
Resolution (Temperature) 0.1°C
Accuracy (Temperature) ±0.5°C
Certifications/Compliance CE marked, ISO 9001, ISO 14001
Material ABS/PC
Display Type LCD
Data Management Details Up to 99 data storage
Outputs RS-232
Temperature Compensation Automatic or Manual
Electrical Requirements 100 to 240V, 50 to 60Hz, 9VDC
Dimensions (L x W x H) 200 x 175 x 52mm
Weight (Metric) 0.6kg

This instrument is suitable for a wide range of applications in various areas, such as the food and beverage industry, as well as water and environmental analysis.

  • Entry-level instrument for measuring pH and mV/ORP
  • Ergonomic and compact design; ABS/PC reinforced housing
  • Fast change over between pH and mV parameters
  • Three-point calibration
  • Separate reference electrode input
  • mV/ORP measuring mode and self-test are combined with useful features such as automated endpoint recognition and calibration with automated buffer recognition
  • Five self-explanatory buttons for simply starting and ending a measurement or for accessing latest calibration data at the press of a button
  • Big, well organized LCD display simultaneously shows readings, temperature, endpoint criteria and various helpful icons and additionally facilitates rapid measurement and calibration
  • Electrode icon provides information on the condition of the electrode
  • Up to 99 data storage
  • RS-232 interface
  • Includes four predefined buffer groups for automatic calibration
  • Separate on/off switch
  • Handy electrode arm automatically included in kit versions


Specifications:
  • pH Range: 0.00 to 14.00
  • pH Resolution: 0.01
  • pH Relative Accuracy: ±0.01
  • mV Range: -1999 to +1999mV
  • mV Resolution: 1
  • mV Relative Accuracy: ±1mV
  • Temperature Range: 0° to 100°C (32° to 212°F)
  • Temperature Resolution: 0.1
  • Temperature Accuracy: ±0.5°C
  • Temperature Compensation: Automatic or Manual
  • Power Supply: 100 to 240V, 50 to 60Hz, 9VDC
  • Dimensions: 200 x 175 x 52mm
  • Weight: 0.6kg
Kits Available:
  • Cat. No. 01-911-301: Meter, operating manual, Quick guide and power adapter
  • Cat. No. 01-911-302: Meter, operating manual, Quick guide, power adapter, LE438 pH electrode, electrode arm, 2X pH 4.01 and 2X pH 7.00 buffer sachets
  • Cat. No. 01-913-996: Meter, operating manual, Quick guide, power adapter, LE420 pH electrode, electrode arm, 2X pH 4.01 and 2X pH 7.00 buffer sachets
  • Cat. No. 01-913-997: Meter, operating manual, Quick guide, power adapter, LE422 pH electrode, electrode arm, 2X pH 4.01 and 2X pH 7.00 buffer sachets
Applications:
  • FEP20-TRIS Kit: Ideal for TRIS buffers.
  • FEP20-Micro Kit: Ideal for small samples.

CE marked, ISO 9001, ISO 14001