Please login to your online account to display your discounted pricing

Mettler Toledo™ FiveEasy Plus™ FEP20 pH Meter

Ideal for anyone who needs an economic and reliable solution for easy pH measurement

Manufacturer: mettler toledo™  51302915

 View all options for this product

Catalog No. 01-911-302

 

Description & Specifications

Specifications

Item Description FiveEasy Plus FEP20 pH Meter: ATC Kit
Includes Meter, operating manual, Quick guide, power adapter, LE438 pH electrode, electrode arm, 2X pH 4.01 and 2X pH 7.00 buffer sachets
Range (pH) 0.00 to 14.00
Resolution (pH) 0.01
Accuracy (pH) ±0.01
Range (mV) -1999 to 1999mV
Resolution (mV) 1mV
Accuracy (mV) ±1mV
Range (Temperature) 0° to 100°C
Resolution (Temperature) 0.1°C
Accuracy (Temperature) ±0.5°C
Display Type LCD
Data Management Details Up to 99 data storage
Outputs RS-232
Certifications/Compliance CE marked, ISO 9001, ISO 14001
Dimensions (L x W x H) 200 x 175 x 52mm
Weight (Metric) 0.6kg
Electrical Requirements 100 to 240V, 50 to 60Hz, 9VDC
Height (Metric) 52mm
Length (Metric) 200mm
Material ABS/PC
Temperature Compensation Automatic or Manual
Temperature Range (English) 32° to +212°F
Temperature Range (Metric) 0° to +100°C
Type FEP20-ATC Meter Kit
Width (Metric) 175mm

This instrument is suitable for a wide range of applications in various areas, such as the food and beverage industry, as well as water and environmental analysis.

  • Entry-level instrument for measuring pH and mV/ORP
  • Ergonomic and compact design; ABS/PC reinforced housing
  • Fast change over between pH and mV parameters
  • Three-point calibration
  • Separate reference electrode input
  • mV/ORP measuring mode and self-test are combined with useful features such as automated endpoint recognition and calibration with automated buffer recognition
  • Five self-explanatory buttons for simply starting and ending a measurement or for accessing latest calibration data at the press of a button
  • Big, well organized LCD display simultaneously shows readings, temperature, endpoint criteria and various helpful icons and additionally facilitates rapid measurement and calibration
  • Electrode icon provides information on the condition of the electrode
  • Up to 99 data storage
  • RS-232 interface
  • Includes four predefined buffer groups for automatic calibration
  • Separate on/off switch
  • Handy electrode arm automatically included in kit versions
 
Specifications:
  • pH Range: 0.00 to 14.00
  • pH Resolution: 0.01
  • pH Relative Accuracy: ±0.01
  • mV Range: -1999 to +1999mV
  • mV Resolution: 1
  • mV Relative Accuracy: ±1mV
  • Temperature Range: 0° to 100°C (32° to 212°F)
  • Temperature Resolution: 0.1
  • Temperature Accuracy: ±0.5°C
  • Temperature Compensation: Automatic or Manual
  • Power Supply: 100 to 240V, 50 to 60Hz, 9VDC
  • Dimensions: 200 x 175 x 52mm
  • Weight: 0.6kg
Kits Available:
  • Cat. No. 01-911-301: Meter, operating manual, Quick guide and power adapter
  • Cat. No. 01-911-302: Meter, operating manual, Quick guide, power adapter, LE438 pH electrode, electrode arm, 2X pH 4.01 and 2X pH 7.00 buffer sachets
  • Cat. No. 01-913-996: Meter, operating manual, Quick guide, power adapter, LE420 pH electrode, electrode arm, 2X pH 4.01 and 2X pH 7.00 buffer sachets
  • Cat. No. 01-913-997: Meter, operating manual, Quick guide, power adapter, LE422 pH electrode, electrode arm, 2X pH 4.01 and 2X pH 7.00 buffer sachets
 Applications:
  • FEP20-TRIS Kit: Ideal for TRIS buffers.
  • FEP20-Micro Kit: Ideal for small samples.