Please login to your online account to display your discounted pricing

Mettler Toledo™ Accessories for FiveEasy™ Meters

For use with FiveEasy pH meters.

$140.88 - $140.88

Specifications

For Use With (Application) pH
View More Specs
Products
Catalog Number Mfr. No. For Use With Type Price Quantity    

01-915-683

 
mettler toledo™
ME51302951
pH meter Electrode Arm Each for $140.88
Description & Specifications

Specifications

For Use With (Application) pH

For use with FiveEasy pH meters.